Ethyl 3-oxo-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylate

Lieferant: Apollo Scientific
Achtung

OR110945-1G
APOSOR110945-1GEA 140 EUR
APOSOR110945-1G
Ethyl 3-oxo-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylate
Ethyl 3-oxo-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylate
Formel: C₁₁H₁₁NO₄
Molecular Weight: 221,209 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11451945
CAS-Nummer: 1038478-70-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR