Methyl (2-nitrophenyl)acetate 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 2-(2-nitrophenyl)acetate
OR110947-5G
OR110947-25G
APOSOR110947-5GEA
23.2
EUR
APOSOR110947-5G
APOSOR110947-25G
Methyl (2-nitrophenyl)acetate 98%
Methyl (2-nitrophenyl)acetate
Formel:
C₉H₉NO₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00968465 CAS-Nummer: 30095-98-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...