Methyl (2-nitrophenyl)acetate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-(2-nitrophenyl)acetate

OR110947-5G OR110947-25G
APOSOR110947-5GEA 23.2 EUR
APOSOR110947-5G APOSOR110947-25G
Methyl (2-nitrophenyl)acetate 98%
Methyl (2-nitrophenyl)acetate
Formel: C₉H₉NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00968465
CAS-Nummer: 30095-98-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR