2-(4-Chlorophenoxy)nicotinonitrile

Lieferant: Apollo Scientific
Achtung

OR110948-1G
APOSOR110948-1GEA 256 EUR
APOSOR110948-1G
2-(4-Chlorophenoxy)nicotinonitrile
2-(4-Chlorophenoxy)nicotinonitrile
Formel: C₁₂H₇ClN₂O
Molecular Weight: 230,65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09929608
CAS-Nummer: 82524-00-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR