meso-Tetraphenylporphin

Lieferant: Apollo Scientific
Achtung

OR11095-250MG
APOSOR11095-250MGEA 21.9 EUR
APOSOR11095-250MG
meso-Tetraphenylporphin
meso-Tetraphenylporphin
Formel: C₄₄H₃₀N₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00011680
CAS-Nummer: 917-23-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR