5-Amino-1-benzofuran-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Aminobenzofuran-2-carboxylic acid

OR110952-1G
APOSOR110952-1GEA 27.7 EUR
APOSOR110952-1G
5-Amino-1-benzofuran-2-carboxylic acid
5-Amino-1-benzofuran-2-carboxylic acid
Formel: C₉H₇NO₃
Molecular Weight: 177.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11505019
CAS-Nummer: 42933-44-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR