N-Methyl-1H-indole-2-carboxamide
Lieferant: Apollo Scientific
|
Achtung
|
OR110959-1G
APOSOR110959-1GEA
180
EUR
APOSOR110959-1G
N-Methyl-1H-indole-2-carboxamide
N-Methyl-1H-indole-2-carboxamide
Formel:
C₁₀H₁₀N₂O Molecular Weight: 174,199 g/mol Schmelzpunkt: 215…217 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03015880 CAS-Nummer: 69808-71-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...