2-(3,4-Dichlorophenyl)acetamide

Lieferant: Apollo Scientific
Achtung

OR110962-1G
APOSOR110962-1GEA 133 EUR
APOSOR110962-1G
2-(3,4-Dichlorophenyl)acetamide
2-(3,4-Dichlorophenyl)acetamide
Formel: C₈H₇Cl₂NO
Molecular Weight: 204,053 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11043647
CAS-Nummer: 868697-78-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR