4-Nitrobenzhydrazid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Nitrobenzoic hydrazide
OR110966-25G
APOSOR110966-25GEA
72.5
EUR
APOSOR110966-25G
4-Nitrobenzhydrazid
4-Nitrobenzhydrazid
Formel:
C₇H₇N₃O₃ Molecular Weight: 181,15 g/mol Schmelzpunkt: 216…218 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007604 CAS-Nummer: 636-97-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...