Boc-3-amino-3-(3-bromophenyl)propionic acid

Lieferant: Apollo Scientific

Synonyme: 3-(3-Bromophenyl)-3-[(tert-butoxycarbonyl)amino]propanoic acid, 3-(3-Bromophenyl)-3-((tert-butoxycarbonyl)amino)propanoic acid

OR110967-1G
APOSOR110967-1GEA 98.6 EUR
APOSOR110967-1G
Boc-3-amino-3-(3-bromophenyl)propionic acid
Boc-3-amino-3-(3-bromophenyl)propionic acid
Formel: C₁₄H₁₈BrNO₄
Molecular Weight: 344,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02090700
CAS-Nummer: 284493-58-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR