2-Chlor-2'-nitroacetanilid

Lieferant: Apollo Scientific
Achtung

OR11097-1G
APOSOR11097-1GEA 21.9 EUR
APOSOR11097-1G
2-Chlor-2'-nitroacetanilid
2-Chlor-2'-nitroacetanilid
Formel: C₈H₇ClN₂O₃
Molecular Weight: 214,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00024201
CAS-Nummer: 10147-70-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR