7-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR110971-1G
APOSOR110971-1GEA 277 EUR
APOSOR110971-1G
7-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid
7-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid
Formel: C₈H₅ClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09991784
CAS-Nummer: 1020038-42-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR