Ethyl-3-tert-butyl-1H-pyrazole-5-carboxylate

Lieferant: Apollo Scientific

Synonyme: 3-tert-Butyl-1H-pyrazole-5-carboxylic acid ethyl ester

OR110974-1G
APOSOR110974-1GEA 0 EUR
APOSOR110974-1G
Ethyl-3-tert-butyl-1H-pyrazole-5-carboxylate
Ethyl-3-tert-butyl-1H-pyrazole-5-carboxylate
Formel: C₁₀H₁₆N₂O₂
Molecular Weight: 196,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173782
CAS-Nummer: 916791-97-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR