N-[2-(4-tert-Butylphenoxy)benzyl]-N-methylamine

Lieferant: Apollo Scientific

OR110979-1G
APOSOR110979-1GEA 248 EUR
APOSOR110979-1G
N-[2-(4-tert-Butylphenoxy)benzyl]-N-methylamine
N-[2-(4-tert-Butylphenoxy)benzyl]-N-methylamine
Formel: C₁₈H₂₃NO
Molecular Weight: 269,381 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12453385
CAS-Nummer: 1095104-40-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR