N-(2,6-Dichlorobenzyl)-N-methylamine

Lieferant: Apollo Scientific
Gefahr

OR110982-1G
APOSOR110982-1GEA 0 EUR
APOSOR110982-1G
N-(2,6-Dichlorobenzyl)-N-methylamine
N-(2,6-Dichlorobenzyl)-N-methylamine
Formel: C₈H₉Cl₂N
Molecular Weight: 190.07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04532390
CAS-Nummer: 15205-19-3
UN: 3265
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR