4-(4-Formylphenoxy)benzonitril

Lieferant: Apollo Scientific

OR110983-1G
APOSOR110983-1GEA 106 EUR
APOSOR110983-1G
4-(4-Formylphenoxy)benzonitril
4-(4-Formylphenoxy)benzonitril
Formel: C₁₄H₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08276813
CAS-Nummer: 90178-71-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR