4-Bromo-1-methyl-1H-pyrazole-3-carbaldehyde oxime

Lieferant: Apollo Scientific

OR110984-1G
APOSOR110984-1GEA 0 EUR
APOSOR110984-1G
4-Bromo-1-methyl-1H-pyrazole-3-carbaldehyde oxime
4-Bromo-1-methyl-1H-pyrazole-3-carbaldehyde oxime
Formel: C₅H₆BrN₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD02090006
CAS-Nummer: 287922-69-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR