Thieno[3,2-e][1,2,4]triazolo[4,3-c]pyrimidine

Lieferant: Apollo Scientific

OR110985-1G
APOSOR110985-1GEA 371 EUR
APOSOR110985-1G
Thieno[3,2-e][1,2,4]triazolo[4,3-c]pyrimidine
Thieno[3,2-e][1,2,4]triazolo[4,3-c]pyrimidine
Formel: C₇H₄N₄S
Molecular Weight: 176,198 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26939174
CAS-Nummer: 18740-36-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR