4,5,6,7-Tetrahydro-1-benzothiophene-2-sulphonamide

Lieferant: Apollo Scientific

OR110986-1G
APOSOR110986-1GEA 445 EUR
APOSOR110986-1G
4,5,6,7-Tetrahydro-1-benzothiophene-2-sulphonamide
4,5,6,7-Tetrahydro-1-benzothiophene-2-sulphonamide
Formel: C₈H₁₁NO₂S₂
Molecular Weight: 217,308 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25958940
CAS-Nummer: 142294-64-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR