1-[4-(2-Methyl-1,3-thiazol-4-yl)phenyl]-5-oxopyrrolidine-3-carboxylic acid

Lieferant: Apollo Scientific

OR110988-1G
APOSOR110988-1GEA 0 EUR
APOSOR110988-1G
1-[4-(2-Methyl-1,3-thiazol-4-yl)phenyl]-5-oxopyrrolidine-3-carboxylic acid
1-[4-(2-Methyl-1,3-thiazol-4-yl)phenyl]-5-oxopyrrolidine-3-carboxylic acid
Formel: C₁₅H₁₄N₂O₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD26939172
CAS-Nummer: 1858257-28-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR