1-(1-Benzothienphen-5-yl)-1H-pyrrole

Lieferant: Apollo Scientific

OR110989-1G
APOSOR110989-1GEA 371 EUR
APOSOR110989-1G
1-(1-Benzothienphen-5-yl)-1H-pyrrole
1-(1-Benzothienphen-5-yl)-1H-pyrrole
Formel: C₁₂H₉NS
Molecular Weight: 199,272 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21102469
CAS-Nummer: 1481442-39-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR