N,4-Dimethyl-1,3-thiazole-5-sulphonamide

Lieferant: Apollo Scientific

OR110990-1G
APOSOR110990-1GEA 296 EUR
APOSOR110990-1G
N,4-Dimethyl-1,3-thiazole-5-sulphonamide
N,4-Dimethyl-1,3-thiazole-5-sulphonamide
Formel: C₅H₈N₂O₂S₂
Molecular Weight: 192,259 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27995613
CAS-Nummer: 1823927-61-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR