Methyl 2-tert-butylimidazo[1,2-a]pyridine-6-carboxylate

Lieferant: Apollo Scientific

OR110992-1G
APOSOR110992-1GEA 322 EUR
APOSOR110992-1G
Methyl 2-tert-butylimidazo[1,2-a]pyridine-6-carboxylate
Methyl 2-tert-butylimidazo[1,2-a]pyridine-6-carboxylate
Formel: C₁₃H₁₆N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD28103336
CAS-Nummer: 1003577-82-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR