2-tert-Butylimidazo[1,2-a]pyridine-6-carboxylic acid

Lieferant: Apollo Scientific

OR110993-1G
APOSOR110993-1GEA 383 EUR
APOSOR110993-1G
2-tert-Butylimidazo[1,2-a]pyridine-6-carboxylic acid
2-tert-Butylimidazo[1,2-a]pyridine-6-carboxylic acid
Formel: C₁₂H₁₄N₂O₂
Molecular Weight: 218,252 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14689252
CAS-Nummer: 1248085-24-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR