1-[2-(4-Nitro-1H-pyrazol-1-yl)ethyl]piperazine

Lieferant: Apollo Scientific

OR110994-1G
APOSOR110994-1GEA 383 EUR
APOSOR110994-1G
1-[2-(4-Nitro-1H-pyrazol-1-yl)ethyl]piperazine
1-[2-(4-Nitro-1H-pyrazol-1-yl)ethyl]piperazine
Formel: C₉H₁₅N₅O₂
Molecular Weight: 225,248 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16076302
CAS-Nummer: 1248142-10-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR