Methyl-3,4,5-trimethoxycinnamate

Lieferant: Apollo Scientific
Achtung

OR11100-1G
APOSOR11100-1GEA 21.9 EUR
APOSOR11100-1G
Methyl-3,4,5-trimethoxycinnamate
Methyl-3,4,5-trimethoxycinnamate
Formel: C₁₃H₁₆O₅
Molecular Weight: 252,27 g/mol
Schmelzpunkt: 108…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03614740
CAS-Nummer: 20329-96-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR