4-(Chloromethyl)-7-methoxy-2H-chromen-2-one

Lieferant: Apollo Scientific
Gefahr

OR111001-1G
APOSOR111001-1GEA 232 EUR
APOSOR111001-1G
4-(Chloromethyl)-7-methoxy-2H-chromen-2-one
4-(Chloromethyl)-7-methoxy-2H-chromen-2-one
Formel: C₁₁H₉ClO₃
Molecular Weight: 224,64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03731259
CAS-Nummer: 41295-55-0
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR