Ethyl-1-pyrimidin-2-ylpiperidine-4-carboxylate 95+%

Lieferant: Apollo Scientific

Synonyme: Ethyl 1-pyrimidin-2-yl-piperidine-4-carboxylate

OR111002-5G
APOSOR111002-5GEA 171 EUR
APOSOR111002-5G
Ethyl-1-pyrimidin-2-ylpiperidine-4-carboxylate 95+%
Ethyl-1-pyrimidin-2-ylpiperidine-4-carboxylate
Formel: C₁₂H₁₇N₃O₂
Molecular Weight: 235,2800 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 111247-60-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR