5-Nitro-2-(tetrahydro-2H-pyran-4-ylmethoxy)pyridine

Lieferant: Apollo Scientific
Achtung

OR111004-1G
APOSOR111004-1GEA 219 EUR
APOSOR111004-1G
5-Nitro-2-(tetrahydro-2H-pyran-4-ylmethoxy)pyridine
5-Nitro-2-(tetrahydro-2H-pyran-4-ylmethoxy)pyridine
Formel: C₁₁H₁₄N₂O₄
MDL: MFCD14666511
CAS-Nummer: 1287218-29-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR