4-(2-Methyl-1,3-thiazol-4-yl)benzonitrile

Lieferant: Apollo Scientific
Achtung

OR111012-10G OR111012-1G OR111012-5G
APOSOR111012-10GEA 458 EUR
APOSOR111012-10G APOSOR111012-1G APOSOR111012-5G
4-(2-Methyl-1,3-thiazol-4-yl)benzonitrile
4-(2-Methyl-1,3-thiazol-4-yl)benzonitrile
Formel: C₁₁H₈N₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD01765038
CAS-Nummer: 127406-79-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR