N-(4-{[(1-Tetrahydro-2H-pyran-4-yl-1H-pyrazol-4-yl)amino]sulphonyl}phenyl)acetamide

Lieferant: Apollo Scientific

OR111013-1G
APOSOR111013-1GEA 493 EUR
APOSOR111013-1G
N-(4-{[(1-Tetrahydro-2H-pyran-4-yl-1H-pyrazol-4-yl)amino]sulphonyl}phenyl)acetamide
N-(4-{[(1-Tetrahydro-2H-pyran-4-yl-1H-pyrazol-4-yl)amino]sulphonyl}phenyl)acetamide
Formel: C₁₆H₂₀N₄O₄S
Molecular Weight: 364,419 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26953774
CAS-Nummer: 1796948-68-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR