N-Methyl-N-[2-(4-methylphenoxy)benzyl]amine

Lieferant: Apollo Scientific

OR111014-1G
APOSOR111014-1GEA 248 EUR
APOSOR111014-1G
N-Methyl-N-[2-(4-methylphenoxy)benzyl]amine
N-Methyl-N-[2-(4-methylphenoxy)benzyl]amine
Formel: C₁₅H₁₇NO
Molecular Weight: 227,302 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11631253
CAS-Nummer: 1095179-72-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR