Ethyl 4-methylthiophene-2-carboxylate

Lieferant: Apollo Scientific

OR111015-1G OR111015-5G
APOSOR111015-1GEA 149 EUR
APOSOR111015-1G APOSOR111015-5G
Ethyl 4-methylthiophene-2-carboxylate
Ethyl 4-methylthiophene-2-carboxylate
Formel: C₈H₁₀O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD12025317
CAS-Nummer: 14282-79-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR