3-(6-Methoxy-2-naphthyl)-1H-pyrazole

Lieferant: Apollo Scientific

OR111017-1G OR111017-250MG
APOSOR111017-1GEA 232 EUR
APOSOR111017-1G APOSOR111017-250MG
3-(6-Methoxy-2-naphthyl)-1H-pyrazole
3-(6-Methoxy-2-naphthyl)-1H-pyrazole
Formel: C₁₄H₁₂N₂O
Molecular Weight: 224.258 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03407411
CAS-Nummer: 258518-56-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR