3-(6-Methoxy-2-naphthyl)-1H-pyrazole
Lieferant: Apollo Scientific
OR111017-1G
OR111017-250MG
APOSOR111017-1GEA
232
EUR
APOSOR111017-1G
APOSOR111017-250MG
3-(6-Methoxy-2-naphthyl)-1H-pyrazole
3-(6-Methoxy-2-naphthyl)-1H-pyrazole
Formel:
C₁₄H₁₂N₂O Molecular Weight: 224.258 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03407411 CAS-Nummer: 258518-56-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...