N-2,3-Dihydro-1-benzofuran-5-yl-2-furamide

Lieferant: Apollo Scientific

OR111022-1G
APOSOR111022-1GEA 306 EUR
APOSOR111022-1G
N-2,3-Dihydro-1-benzofuran-5-yl-2-furamide
N-2,3-Dihydro-1-benzofuran-5-yl-2-furamide
Formel: C₁₃H₁₁NO₃
Molecular Weight: 229,231 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11823250
CAS-Nummer: 1090579-35-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR