1-Methyl-1H-indole-2-carbonitrile
Lieferant: Apollo Scientific
|
Achtung
|
OR111023-1G
APOSOR111023-1GEA
0
EUR
APOSOR111023-1G
1-Methyl-1H-indole-2-carbonitrile
1-Methyl-1H-indole-2-carbonitrile
Formel:
C₁₀H₈N₂ Schmelzpunkt: 67…69 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09263865 CAS-Nummer: 60680-97-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...