(2-Acetyl-4-methylphenoxy)acetic acid

Lieferant: Apollo Scientific

OR111024-1G
APOSOR111024-1GEA 272 EUR
APOSOR111024-1G
(2-Acetyl-4-methylphenoxy)acetic acid
(2-Acetyl-4-methylphenoxy)acetic acid
Formel: C₁₁H₁₂O₄
Molecular Weight: 208,211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03225657
CAS-Nummer: 88521-62-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR