5-Nitro-1-benzofuran

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Nitrobenzofuran

OR111026-1G
APOSOR111026-1GEA 219 EUR
APOSOR111026-1G
5-Nitro-1-benzofuran
5-Nitro-1-benzofuran
Formel: C₈H₅NO₃
Molecular Weight: 163,13 g/mol
Schmelzpunkt: 110…112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00456542
CAS-Nummer: 18761-31-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR