4-(2,4-Dichloro-6-methylphenyl)-3-thiosemicarbazide
Lieferant: Apollo Scientific
OR111027-1G
APOSOR111027-1GEA
296
EUR
APOSOR111027-1G
4-(2,4-Dichloro-6-methylphenyl)-3-thiosemicarbazide
4-(2,4-Dichloro-6-methylphenyl)-3-thiosemicarbazide
Formel:
C₈H₉Cl₂N₃S Molecular Weight: 250,148 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28148228 CAS-Nummer: 1823624-22-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...