6-Nitroimidazo[1,2-a]pyridin Hydrochlorid

Lieferant: Apollo Scientific

OR111033-1G
APOSOR111033-1GEA 0 EUR
APOSOR111033-1G
6-Nitroimidazo[1,2-a]pyridin Hydrochlorid
6-Nitroimidazo[1,2-a]pyridin Hydrochlorid
Formel: C₇H₆ClN₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09800971
CAS-Nummer: 957120-37-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR