2-Phenoxypyridin-3-amine

Lieferant: Apollo Scientific

OR111037-1G
APOSOR111037-1GEA 386 EUR
APOSOR111037-1G
2-Phenoxypyridin-3-amine
2-Phenoxypyridin-3-amine
Formel: C₁₁H₁₀N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD09738154
CAS-Nummer: 28664-62-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR