3-[(2,6-Dimethylmorpholin-4-yl)methyl]benzonitrile
Lieferant: Apollo Scientific
OR111038-1G
APOSOR111038-1GEA
0
EUR
APOSOR111038-1G
3-[(2,6-Dimethylmorpholin-4-yl)methyl]benzonitrile
3-[(2,6-Dimethylmorpholin-4-yl)methyl]benzonitrile
Formel:
C₁₄H₁₈N₂O Molecular Weight: 230,306 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09928897 CAS-Nummer: 1016709-98-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...