N-(4-Chlorophenyl)-2,6-dimethylmorpholine-4-carboxamide
Lieferant: Apollo Scientific
OR111039-1G
APOSOR111039-1GEA
0
EUR
APOSOR111039-1G
N-(4-Chlorophenyl)-2,6-dimethylmorpholine-4-carboxamide
N-(4-Chlorophenyl)-2,6-dimethylmorpholine-4-carboxamide
Formel:
C₁₃H₁₇ClN₂O₂ Molecular Weight: 268,739 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00023340 CAS-Nummer: 77280-30-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...