6-Chloro-2-methyl-2H-1,4-benzoxazin-3(4H)-one
Lieferant: Apollo Scientific
Synonyme:
6-Chloro-2-methyl-2,4-dihydro-1,4-benzoxazin-3-one
OR111040-5G
APOSOR111040-5GEA
0
EUR
APOSOR111040-5G
6-Chloro-2-methyl-2H-1,4-benzoxazin-3(4H)-one
6-Chloro-2-methyl-2H-1,4-benzoxazin-3(4H)-one
Formel:
C₉H₈ClNO₂ Molecular Weight: 197.62 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01744501 CAS-Nummer: 5791-00-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...