N-(4-Chloro-2,5-dimethoxyphenyl)-N'-(1-methyl-1H-indazol-3-yl)thiourea

Lieferant: Apollo Scientific

OR111050-1G
APOSOR111050-1GEA 0 EUR
APOSOR111050-1G
N-(4-Chloro-2,5-dimethoxyphenyl)-N'-(1-methyl-1H-indazol-3-yl)thiourea
N-(4-Chloro-2,5-dimethoxyphenyl)-N'-(1-methyl-1H-indazol-3-yl)thiourea
Formel: C₁₇H₁₇ClN₄O₂S
Molecular Weight: 376,86 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28148248
CAS-Nummer: 1858241-71-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR