N-(2,4-Dichloro-6-methylphenyl)-N'-(1-methyl-1H-indazol-3-yl)thiourea

Lieferant: Apollo Scientific

OR111051-1G
APOSOR111051-1GEA 0 EUR
APOSOR111051-1G
N-(2,4-Dichloro-6-methylphenyl)-N'-(1-methyl-1H-indazol-3-yl)thiourea
N-(2,4-Dichloro-6-methylphenyl)-N'-(1-methyl-1H-indazol-3-yl)thiourea
Formel: C₁₆H₁₄Cl₂N₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD28148249
CAS-Nummer: 1858255-25-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR