4-Amino-N-(1-tetrahydro-2H-pyran-4-yl-1H-pyrazol-4-yl)benzenesulphonamide

Lieferant: Apollo Scientific

OR111058-1G
APOSOR111058-1GEA 493 EUR
APOSOR111058-1G
4-Amino-N-(1-tetrahydro-2H-pyran-4-yl-1H-pyrazol-4-yl)benzenesulphonamide
4-Amino-N-(1-tetrahydro-2H-pyran-4-yl-1H-pyrazol-4-yl)benzenesulphonamide
Formel: C₁₄H₁₈N₄O₃S
Molecular Weight: 322,383 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28148230
CAS-Nummer: 1858251-36-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR