N-Pyridin-3-yl-2-(tetrahydro-2H-pyran-4-ylcarbonyl)hydrazinecarbothioamide

Lieferant: Apollo Scientific

OR111059-1G
APOSOR111059-1GEA 310 EUR
APOSOR111059-1G
N-Pyridin-3-yl-2-(tetrahydro-2H-pyran-4-ylcarbonyl)hydrazinecarbothioamide
N-Pyridin-3-yl-2-(tetrahydro-2H-pyran-4-ylcarbonyl)hydrazinecarbothioamide
Formel: C₁₂H₁₆N₄O₂S
Molecular Weight: 280,346 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28148235
CAS-Nummer: 1858249-71-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR