4-Pyridin-3-yl-5-tetrahydro-2H-pyran-4-yl-4H-1,2,4-triazole-3-thiol

Lieferant: Apollo Scientific

OR111060-1G
APOSOR111060-1GEA 371 EUR
APOSOR111060-1G
4-Pyridin-3-yl-5-tetrahydro-2H-pyran-4-yl-4H-1,2,4-triazole-3-thiol
4-Pyridin-3-yl-5-tetrahydro-2H-pyran-4-yl-4H-1,2,4-triazole-3-thiol
Formel: C₁₂H₁₄N₄OS
Molecular Weight: 262,331 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28148234
CAS-Nummer: 1858250-40-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR